missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Cytidine-5'-diphosphocholine sodium salt

Catalog No. p-7042884
Change view
Click to view available options
Quantity:
10 g
25 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6416109 10 g
AAJ6416114 25 g
AAJ6416118 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ6416109 Supplier Thermo Scientific Chemicals Supplier No. J6416109
Only null left

Intermediate in the synthesis of phosphatidylcholine; also a phospholipase inhibitor | CAS: 33818-15-4 | C14H25N4NaO11P2 | 510.31 g/mol

Cytidine-5'-diphosphocholine sodium salt hydrate acts as a neuroprotective agent in situations of hypoxia and ischemia. It is used as biosynthetic intermediate of membrane phospholipids. It is a cerebral vasodilator and inhibits PLA2 activation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cytidine-5′-diphosphocholine sodium salt hydrate acts as a neuroprotective agent in situations of hypoxia and ischemia. It is used as biosynthetic intermediate of membrane phospholipids. It is a cerebral vasodilator and inhibits PLA2 activation.

Solubility
Soluble in water (100 mg/ml).

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 33818-15-4
Molecular Formula C14H25N4NaO11P2
Molecular Weight (g/mol) 510.31
MDL Number MFCD00150508
InChI Key YWAFNFGRBBBSPD-OCMLZEEQSA-M
Synonym citicolinesodium, cytidine 5'-diphosphocholine sodium, cdp-choline sodium, citicoline, sodium, cdp-choline, sodium, cytidine 5'-diphosphocholine sodium salt, cytidine 5'-diphosphocholine sodium salt dihydrate, cytidine-5'-diphosphocholine monosodium salt, sodium 2s,3r,4s,5s-5-4-amino-2-oxo-1-pyrimidinyl-3,4-dihydroxy-2-oxolanyl methoxy-oxidophosphoryl 2-trimethylammonio ethyl phosphate, sodium 2s,3r,4s,5s-5-4-azanyl-2-oxidanylidene-pyrimidin-1-yl-3,4-bis oxidanyl oxolan-2-yl methoxy-oxidanidyl-phosphoryl 2-trimethylazaniumyl ethyl phosphate
PubChem CID 46738085
IUPAC Name sodium;[[(2S,3R,4S,5S)-5-(4-amino-2-oxopyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-oxidophosphoryl] 2-(trimethylazaniumyl)ethyl phosphate
SMILES [Na+].C[N+](C)(C)CCOP([O-])(=O)OP([O-])(=O)OC[C@H]1O[C@H]([C@H](O)[C@@H]1O)N1C=CC(N)=NC1=O

Specifications

Color White
Quantity 10 g
Beilstein 4110040
Solubility Information Soluble in water (100mg/ml).
Formula Weight 510.31 (Anhydrous)
Physical Form Powder
Chemical Name or Material Cytidine-5'-diphosphocholine sodium salt hydrate
EINECSNumber 251-689-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only