missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytidine 5'-Diphosphate Trisodium Salt Hydrate 95.0+%, TCI America™
Specifications
Specifications
| Color | White |
| Quantity | 10 mg |
| Formula Weight | 469.12 |
| Percent Purity | ≥95.0% (HPLC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Cytidine 5′-Diphosphate Trisodium Salt Hydrate |
Safety and Handling
| TSCA | No |
| Recommended Storage | Refrigerator |