missing translation for 'onlineSavingsMsg'
Learn More

Cystamine Dihydrochloride, 97%

Catalog No. p-3736087
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC111770250 25 g Plastic bottle
AC111771000 100 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC111770250 Supplier Thermo Scientific Chemicals Supplier No. 111770250
Only null left

CAS: 56-17-7 | C4H12N2S2·2HCl | 225.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 56-17-7
Molecular Formula C4H12N2S2·2HCl
Molecular Weight (g/mol) 225.2
MDL Number MFCD00012905
InChI Key YUFRRMZSSPQMOS-UHFFFAOYSA-N
Synonym cystamine dihydrochloride, cystamin dihydrochloride, ethanamine, 2,2'-dithiobis-, dihydrochloride, usaf cb-34, 2,2'-disulfanediyldiethanamine dihydrochloride, 2-aminoethyl disulfide dihydrochloride, bis 2-aminoethyl disulfide dihydrochloride, cystamine 2hcl, 2,2'-dithiodi ethylammonium dichloride, decarboxycystine dihydrochloride
PubChem CID 5941
IUPAC Name 2-(2-aminoethyldisulfanyl)ethanamine;dihydrochloride
SMILES C(CSSCCN)N.Cl.Cl

Specifications

Melting Point 220°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (Argentometry)
Packaging Plastic bottle
Linear Formula S2(CH2CH2NH2)2·2HCl
Quantity 25 g
Beilstein 04, 287
Merck Index 15, 2773
Solubility Information Solubility in water: soluble. Other solubilities: slightly soluble in acetone,ethanol
Formula Weight 225.2
Percent Purity 97%
Physical Form Crystalline Powder, Crystals or Chunks
Chemical Name or Material Cystamine dihydrochloride
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 200-260-7

RUO – Research Use Only