missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CyQUANT™ Cell Proliferation Assays

Catalog No. c35007
Encompass
Change view
Click to view available options
Cell Type:
Cell Lysis Buffer
Direct Cell
Direct Red Cell
For cells in culture
NF Cell
Quantity:
10 Microplates
100 Microplates
1000 Assays
200 Assays
50 mL
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Cell Type Quantity
C35007 NF Cell 200 Assays
C35011 Direct Cell 10 Microplates
C35013 Direct Red Cell 10 Microplates
C35012 Direct Cell 100 Microplates
C7026 For cells in culture 1000 Assays
C35006 NF Cell 1000 Assays
C7027 Cell Lysis Buffer 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. C35007 Supplier Invitrogen™ Supplier No. C35007

CyQUANT cell proliferation assays are fast and highly sensitive fluorescence methods for counting cells to assess cell growth and cytotoxicity in microplate format. These assays use fluorescent nucleic-acid-binding dyes to measure cell number based on DNA content.

Since the amount of DNA in cells is highly regulated, methods to detect DNA content offer improved accuracy over metabolism-based cellular proliferation or cytotoxicity assays, which can be affected by cell changes that are unrelated to cell number. CyQUANT cell proliferation assays use cell-permeant, DNA-binding dyes that enhance fluorescence when bound to nucleic acids to measure cellular DNA content. The resulting fluorescent signal can be compared to standards to determine cell number, enabling fast and sensitive microplate assays to measure cell proliferation and cytotoxicity.

Choose from a range of CyQUANT cell proliferation assays based on experimental needs and workflows.

The CyQUANT and CyQUANT NF (No Freeze) cell proliferation assays provide cell proliferation measurements based on the total cell count, including both live and dead cells. The CyQUANT Direct Cell Proliferation Assay only detects live, healthy cells by using a masking dye that blocks staining of dead cells with compromised membranes, providing measurement of both cellular proliferation and cytotoxicity.

CyQUANT Cell Proliferation Assay

The CyQUANT Cell Proliferation Assay (Cat. No. C7026) enables precise DNA measurement for total cell count using the green-fluorescent CyQUANT GR dye and allows for the convenient freezing and storage of samples for up to four weeks. Easily compare multiple samples with minimal hands-on time by freezing, lysing, and reading fluorescence on a standard microplate reader.

Features of the CyQUANT Cell Proliferation Assay include:

  • Direct DNA measurement

    —utilizes the green-fluorescent CyQUANT GR dye (ex/em 480/520 nm), a proprietary dye with strong fluorescence enhancement when bound to nucleic acids, for accurate measurement of DNA content to count both live and dead cells
  • Broad linear range

    —quantify 50 to 50,000 cells per well without relying on metabolic activity
  • Quickly compare samples

    —compare multiple samples in a microplate format with minimal hands-on time; simply remove media, freeze and lyse cells, then read fluorescence on a standard microplate reader with a fluorescein filter
  • Convenient sample storage

    —freeze and store time course samples for up to four weeks, enabling batch analysis and direct comparison of samples taken at different time points
  • Comprehensive kit

    —includes the CyQUANT GR dye, cell lysis buffer, and bacteriophage λ DNA as a reference standard; extra CyQUANT Cell Lysis Buffer available separately (Cat. No. C7027)

CyQUANT NF (No Freeze) Cell Proliferation Assay

The CyQUANT NF (No Freeze) Cell Proliferation Assay (Cat. Nos. C35006 and C35007) offers a streamlined protocol for total cell count by eliminating the freeze-thaw cell lysis step of the original CyQUANT Cell Proliferation Assay using a cell-permeant DNA-binding dye and a plasma membrane permeabilization/dye delivery reagent. This sensitive and rapid cell counting assay provides a wide detection range and is completed in just one hour without the need for cell lysis.

Features of the CyQUANT NF Cell Proliferation Assay include:

  • Streamlined protocol

    —eliminates the freeze-thaw cell lysis step using the cell-permeant and green-fluorescent CyQUANT NF DNA-binding dye (ex/em 480/520 nm) in combination with a plasma membrane permeabilization reagent for accurate measurement of DNA content to count both live and dead cells
  • Wide detection range

    —linear detection range from 100 to 20,000 cells per well in a 96-well microplate
  • Sensitive and rapid assay

    —more sensitive than MTT or alamarBlue™ assays, complete proliferation protocol in just one hour
  • No cell lysis required

    —eliminates the need for cell lysis, long incubations, radioactivity, or stain removal

CyQUANT Direct Cell Proliferation Assay

The CyQUANT Direct Cell Proliferation Assay (Cat. Nos. C35011, C35012, and C35013) provides a simple, no-wash, add-and-read workflow ideal for high-throughput screening cellular proliferation and cytotoxicity assays. This assay uses a green- or red-fluorescent cell-permeant DNA-binding probe that enters all cells and a cell-impermeant background suppressor that blocks staining in dead cells or cells with compromised membranes, ensuring that only live, healthy cells are stained. The assay measures changes in cell proliferation as well as cytotoxicity to allow for accurate assessment of cell growth, viability, and compound toxicity.

Features of the CyQUANT Direct Cell Proliferation Assay include:

  • Dual measurement

    —assesses both cell proliferation and membrane integrity, staining only live, viable cells
  • Convenient workflow

    —no washes, cell lysis, temperature equilibrations, or radioactivity required; ideal for high-throughput screening
  • Two fluorescent options

    —available in green CyQUANT Direct (ex/em 508/527 nm) and red CyQUANT Direct Red (ex/em 622/645 nm) for multiplexing with other fluorescent dyes or proteins; can switch between HTS and HCS plate readers if multiplex assays require different platforms
  • Wide detection range

    —linear detection from less than 100 to 20,000 cells per well in most cell types
  • Rapid and easy protocol

    —no-wash, homogeneous format with a fast proliferation protocol; simply mix the probe and suppressor, add to cells, incubate for one hour, and measure fluorescence
  • Stable signal

    —fluorescent signal remains stable for several hours, providing workflow flexibility
  • Multiplexing capability

    —compatible with spectrally distinct fluorescent probes or luminescence assays because cells are not lysed, allowing use with different platforms
  • Correlation with metabolic assays

    —excellent concordance with metabolism- or energy (ATP)-based cell health assays for reliable comparison of historical data

Specifications

Cell Type NF Cell
Content And Storage •CyQUANT™ NF dye reagent
•Dye delivery reagent
•5X HBSS buffer (Hank’s balanced salt solution)

It is possible to observe precipitate in HBSS buffer. The precipitate does not have an impact on the use of this kit, nor the results that are gathered from its use.
Detection Method Fluorescence
For Use With (Application) Proliferation Assay
For Use With (Equipment) Microplate Reader, HTS Reader
Product Type Cell Proliferation Assay
Dye Type Other Label(s) or Dye(s)
Format 384-well plate, 96-well plate
Product Line CyQUANT
Quantity 200 Assays
Shipping Condition Room Temperature
Excitation/Emission 480/520 nm
Show More Show Less
What is the difference between the original CyQuant assay and the CyQuant NF assay?

The original CyQUANT assay provides sensitive detection of cells over a 1000-fold linear dynamic range. In this assay, a freeze-thaw cell lysis step is required to facilitate the interaction of the CyQUANT GR dye with DNA. The CyQUANT NF assay avoids this freeze-thaw step by using a DNA binding dye in combination with a plasma membrane permeabilization reagent. The CyQUANT NF protocol requires only aspiration of growth medium (for adherent cells), replacement with dye binding solution, incubation for 30-60 minutes, and then measurement of fluorescence in a microplate reader. The assay is designed to produce a linear analytical response from at least 100-20,000 cells per well in most cell lines in a 96-well microplate.

Do you offer any products to measure neuronal cell health?

PrestoBlue Cell Viability Stain and CyQUANT Cell Proliferation Assay Kit can be used. We also offer a Neurite Outgrowth Staining Kit (Cat. No. A15001). More information about our different assays for neuronal cell health can be found here (https://www.thermofisher.com/us/en/home/life-science/cell-analysis/neuroscience/neuronal-cell-health-assays.html).

For Research Use Only. Not for use in diagnostic procedures.