missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CyQUANT™ MTT and XTT Cell Viability Assays

Catalog No. p-200110075
Encompass
Change view
Click to view available options
Dye Type:
MTT
XTT
Emission Wavelength Range:
450, 660
570
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dye Type Emission Wavelength Range
V13154 MTT 570
X12223 XTT 450, 660
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. V13154 Supplier Invitrogen™ Supplier No. V13154

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CyQUANT MTT and XTT Cell Viability assays are colorimetric microplate assays for accurate assessment of cell health, proliferation, and cytotoxicity. CyQUANT MTT is a well-established assay, while CyQUANT XTT offers improved performance and continuous measurement without cell lysis or solubilization.

CyQUANT MTT and XTT Cell Viability assays are colorimetric microplate assays used to measure the metabolic activity of cells, which is an indicator of cell viability, proliferation, and cytotoxicity. CyQUANT MTT and XTT function as cell health indicators by using the cellular redox potential of live cells to convert a tetrazolium-based salt reagent, MTT or XTT, to a brightly colored formazan product. The absorbance of the formazan can be measured using a microplate reader and correlates with the number of metabolically active cells to quantitively measure cell viability and proliferation.

Measuring cell viability is essential for assessing cell health, determining genotoxicity, and evaluating anti-cancer drugs. Colorimetric indicators such as MTT and XTT offer rapid, cost-effective, and consistent methods for quantitatively determining mammalian cell viability using microplate readers. The CyQUANT MTT assay is a well-established and popular cell viability assay, while the CyQUANT XTT assay provides a high dynamic range and low variability. In addition to improved performance, the CyQUANT XTT assay offers continuous measurement without the need for cell lysis or solubilization.

Features of the CyQUANT MTT Cell Viability Assay

  • Complete and optimized MTT assay kit—includes all reagents needed for colorimetric detection of viable mammalian cells
  • Trusted MTT reagent—uses the well-established MTT reagent to measure cellular redox potential
  • Configured to perform 1,000 tests using a 96-well microplate

Principle of the CyQUANT MTT Cell Viability Assay

The CyQUANT MTT assay is based on the reduction of the water-soluble, yellow tetrazolium salt MTT (3-(4,5-dimethylthiazol-2-yl)-2,5-diphenyltetrazolium bromide) to insoluble, purple formazan crystals by metabolically active cells. The redox potential in the mitochondria of active mammalian cells reduces MTT to a strongly pigmented formazan product. After solubilization, the absorbance of the formazan can be measured on a microplate reader, which correlates with the number of metabolically active cells.

Protocol for the CyQUANT MTT Cell Viability Assay

Prepare the MTT reagent and incubate cells for a specific period, during which viable cells convert MTT into insoluble purple formazan product. The formazan crystals are then solubilized with the included SDS (sodium dodecyl sulfate) reagent or DMSO, and the absorbance is measured using a microplate reader at 570 nm.

Features of the CyQUANT XTT Cell Viability Assay

  • High reproducibility—absorbance-based viability assay that measures redox potential in live cells with low variability
  • Improved dynamic range and sensitivity—enhanced performance compared to other colorimetric viability assays
  • Easy mix-and-read format—configured for one 96-well plate at a time
  • Non-toxic and continuous—allows multiple time points to be collected without cell lysis
  • Quick and simple—no solubilization step required, making it faster and easier to perform than the MTT assay
  • Complete XTT assay kit—includes all the materials needed for ten 96-well plates

Principle of the CyQUANT XTT Cell Viability Assay

The CyQUANT XTT assay relies on the reduction of the water-soluble, yellow tetrazolium salt XTT (2,3-bis-(2-methoxy-4-nitro-5-sulfophenyl)-2H-tetrazolium-5-carboxanilide) to a water-soluble orange formazan product by metabolically active cells. Unlike MTT, XTT does not form insoluble crystals and produces a soluble formazan product that does not require solubilization. Since no cell lysis or solubilization are required for detection, the XTT assay can be used as a continuous assay, and multiple time points can be analyzed using an absorbance-based microplate reader.

In addition to the XTT Reagent, the CyQUANT XTT Cell Viability Assay kit includes electron coupling reagent to improve the dynamic range of the assay. The sensitivity and consistency of the assay is significantly increased when the electron coupling reagent is used with the CyQUANT XTT assay kit.

Protocol for the CyQUANT XTT Cell Viability Assay

For each 96-well plate, one bottle of XTT reagent and one tube of electron coupling reagent are thawed and mixed to create a stock solution, then added to cells. Cells are incubated for 2–4 hours, and metabolically active cells reduce XTT to form a water-soluble orange formazan dye. The absorbance of the dye is then measured using a microplate reader at 450 nm and 650 nm.

The XTT/electron coupling reagent stock solution should be used promptly after mixing, as extended room temperature storage or freeze/thaw cycles reduce assay performance and sensitivity. To ensure optimal performance and reproducibility, the CyQUANT XTT assay kit consists of separate XTT and electron coupling reagents that are mixed just prior to use.

Order Info

Shipping Condition: Room temperature. The kit provides a sufficient amount of material for approximately 1000 assays.

Specifications

Cell Type Mammalian Cells
Content And Storage Store at 2°C to 8°C and protect from light.
Detection Method Absorbance
For Use With (Application) Viability Assay, Proliferation Assay, Cytotoxicity Assay
For Use With (Equipment) Microplate Reader
Product Type Cell Viability Assay
Dye Type MTT
Format 96-well plate
Product Line CyQUANT
Quantity 1000 Assays
Shipping Condition Room Temperature
Emission Wavelength Range 570
Show More Show Less
MTT cell proliferation plate assays require cellular metabolism to modify the reagent and thus, only live cells will be counted. Is this true for CyQUANT Direct Cell Proliferation Assay, too?

This is true for MTT (as well as XTT, AlamarBlue Cell Viability Reagent, and PrestoBlue Cell Viability Reagent). CyQUANT Direct will also only count live cells, but for a different reason. The dye in it is a green-fluorescent nucleic acid stain, which will bind to DNA in all cells, live or dead, without the need for cellular metabolism. However, there is a non-cell-permeable quenching reagent in the kit which will both quench extracellular fluorescence (and thus this is a no-wash assay) AND will quench the fluorescence in dead cells (but not live cells).

MTT cell proliferation assays require cellular metabolism to turn over the reagent, and thus only live cells will be counted. Is this true for CyQUANT Direct as well?

This is true for MTT (as well as XTT, AlamarBlue Cell Viability Reagent, and PrestoBlue Cell Viability Reagent). CyQUANT Direct will also only count live cells, but for a different reason. The dye is a green-fluorescent nucleic acid stain, which will bind to DNA in all cells, live or dead, without the need for cellular metabolism. However, there is a cell impermeant quenching reagent in the kit which will quench both extracellular fluorescence (and thus this is a no-wash assay) and the fluorescence in dead cells (but not live cells).

I want to use MTT to determine cell proliferation. Do I really need a standard curve? Will the assay tell me percent of live versus dead cells?

MTT is reduced only by live cells and the rate of dye reduction may vary from cell to cell based on their life cycle stage, age, health and other factors. This assay would not reveal the number of dead cells or a ratio of live to dead cells. A standard curve is recommended to determine an approximate number of live cells per sample.

To obtain a ratio of live to dead cells, use a two color fluorescent assay, such as LIVE/DEAD Viability/Cytotoxicity Kit (Cat. No. L3224) or differences in intensity of a single fluorescent color, using a flow cytometer and the LIVE/DEAD Fixable Kits.

For Research Use Only. Not for use in diagnostic procedures.