missing translation for 'onlineSavingsMsg'
Learn More

Cyproheptadine Hydrochloride Sesquihydrate 98.0+%, TCI America™

Catalog No. C32185G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C32185G 5 g
C321825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C32185G Supplier TCI America Supplier No. C32185G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 41354-29-4
Molecular Formula C42H50Cl2N2O3
Molecular Weight (g/mol) 701.77
MDL Number MFCD27967225,MFCD00012538,MFCD00012538,MFCD27967225,MFCD00242817
InChI Key ZEAUHIZSRUAMQG-UHFFFAOYSA-N
Synonym 4-(5H-Dibenzo[a,d]cyclohepten-5-ylidene)-1-methylpiperidine Hydrochloride
PubChem CID 74764338
ChEBI CHEBI:650917
IUPAC Name dihydrogen bis(1-methyl-4-{tricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,9,11,13-heptaen-2-ylidene}piperidine) trihydrate dichloride
SMILES [H+].[H+].O.O.O.[Cl-].[Cl-].CN1CCC(CC1)=C1C2=CC=CC=C2C=CC2=CC=CC=C12.CN1CCC(CC1)=C1C2=CC=CC=C2C=CC2=CC=CC=C12

Specifications

Color White-Yellow
Quantity 5 g
UN Number 2811
Formula Weight 323.86
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Cyproheptadine Hydrochloride Sesquihydrate
RTECSNumber TM7050000
TSCA No