missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ pCMV-Cypridina Luc Vector for Luciferase Assays

Catalog No. PI16150
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI16150 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI16150 Supplier Thermo Scientific™ Supplier No. LSG16150

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Drive Cypridina luciferase expression as a reporter for gene regulation studies using vectors with a multiple cloning site (MCS) or control promoter (CMV or TK).

The Thermo Scientific™ pMCS-Cypridina Luc Vector is a multiple cloning site plasmid designed to accept a promoter sequence for study of gene regulation using the naturally secreting Cypridina luciferase reporter.

The Cypridina Luc Vectors contain a gene cloned from the marine ostracod, Cypridina noctiluca. The gene encodes a naturally-secreted bioluminescent Cypridina luciferase (62kDa), which enables measurement of the reporter activity in media (for real-time assays) and in cell lysates. The pMCS vector contains a multiple cloning site for cloning a promoter to study its regulatory potential. The pCMV and pTK vectors have the luciferase gene under the CMV (Cytomegalovirus) promoter and Herpes Simplex Virus (HSV) thymidine kinase (TK) promoter, respectively. These constitutive expression vectors can be used as normalization controls to account for experimental variation in combination with other reporters.

Highlights:

  • Naturally-secreting Cypridina luciferase gene, optimized for high expression in mammalian systems
  • Multiple cloning site (MCS) provides versatility for transfer of regulatory elements from one plasmid to another
  • Transcription termination site (Ter), Lac operator (Lac O1), and transcriptional pause site (TPS) used to minimize background by reducing transcriptional read-through
  • Both puromycin (Pur) and ampicillin (Amp) markers for drug selection in mammalian and bacterial cells, respectively
  • High-copy pUC bacterial DNA replication origin
  • Two control vectors available with strong (CMV) or weak (TK) constitutive promoters for co-transfection and normalization

Specifications

Description pCMV-Cypridina Luc Vector
Type Luc Vector
Cloning Method Restriction Enzyme/MCS
Delivery Type Transfection
Quantity 10 μg
Content And Storage Store between -70°C and -20°C.
Reporter Gene Cypridina Luciferase
Selection Agent (Eukaryotic) Puromycin
Product Line Pierce
For Use With (Application) Reporter Assays
Promoter CMV
Show More Show Less
What options do you offer for luciferase vectors?

There are three vectors available for each expressed luciferase; pMCS, pCMV, and pTK. We recommend using pMCS to measure the activity of the reporter, and pCMV, and pTK as positive control vectors. For high level constitutive expression of Luciferase, we recommend using the CMV (Cytomegalovirus) promoter vector and for low level constitutive expression of luciferase, we recommend using the pTK (thymidine kinase) promoter vector.

What is the stability of the secreted luciferase?

Gaussia, Gaussia-Dura, and Cypridina Luciferase are stable in the culture media for greater than 24 hours. Generally, after transfection of CMV driven constructs, we see ever increasing secreted signal between 24 and 72 hours.

How quickly is luciferase secreted?

The earliest detectable signal will depend on the strength of the promoter. In general, with a strong promoter (such as CMV), significant signal over background can be seen in as little as 20 minutes. However, with a weaker inducible promoter, significant signal over background may take 1-2 hours after induction.

Do you still offer quantitative IP (qIP) Luciferase assay kits?

Our qIP Luciferase assay kits have been discontinued but we do carry epitope-tagged (HA or c-Myc) and Tluc (TurboLuc luciferase)-tagged mammalian expression vectors, and qIP protein interaction assay reagents for these assay kits.

For Research Use Only. Not for use in diagnostic procedures.