missing translation for 'onlineSavingsMsg'
Learn More

Cytiva CypHer5E NHS Esters

Catalog No. 09928189
Change view
Click to view available options
Quantity:
1 mg
5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
09928189 5 mg
09928188 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 09928189 Supplier Cytiva Supplier No. PA15405
Only null left

A red-excitable, pH-sensitive cyanine dye derivative, reporting receptors movement from the cell surface into acidic endosomes upon agonist stimulation.

CypHer5E is a flexible, generic fluorophore that can be used to study most classes of cell surface receptors which internalize into the endocytic pathway when activated by a ligand.

  • Generic technology can be coupled to any receptor or epitope tag antibody.
  • Monitors any neutral to acidic event.
  • Simple fluorescence intensity increase.
  • Classic CyDye profile of two heterocycles linked by a polymethine bridge, and contains secondary nitrogen atoms in one or both heterocyclic rings.
  • CypHer5E has additional sulfonate groups to enhance its solubility in aqueous solutions.
  • Increased labelling efficiency and reduces adverse dye-dye interactions.

CypHer5E reports ligand-mediated receptor internalization by exploiting pH differences that occur along the endocytic pathway. CypHer5E is a red-excitable, pH-sensitive cyanine dye derivative that is minimally fluorescent at a basic pH and maximally fluorescent at an acidic pH. It is therefore well suited to report the movement of a receptor from the cell surface into acidic endosomes upon agonist stimulation.

Specifications

Formulation 100% CypHer 5E
For Use With (Application) This reagent has been shown to be suitable for live cell receptor internalization studies using the IN Cell Analyzer 3000, IN Cell Analyzer 1000, and confocal microscopes. 0.5 mL of stock reagent will be sufficient for up to 500 wells.
Quantity 5 mg
Product Type CypHer5E NHS Ester
Solubility Easily soluble in methanol; Very slightly soluble in cold water and hot water.
Color Blue
Emission 663