missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CYP4Z1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA515052
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA515052 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA515052 Supplier Invitrogen™ Supplier No. PA515052
Only null left

Rabbit Polyclonal Antibody

CYP4Z1 Polyclonal Antibody for Western Blot

The cytochrome P450 (P450) superfamily of enzymes is one of three enzyme systems which metabolize the fatty acid arachadonic acid (AA) to regulators of vascular tone. P450 enzymes are monooxygenase enzymes which require several co-factors such as nicotinamide adenine dinucleotide phosphate (NADPH) and P450 reductase. There are over 200 known genes which encode P450s. Epoxygenases are those P450s which metabolize AA to epoxyeicosatrienoic acid (EETs) and omega-hydroxylases are those P450s which produce 19- and 20-hydroxyeicosatetraenoic acids (19- and 20-HETE). As well as fatty acid metabolism, P450s also metabolize many drugs and toxins. Cytochrome P450 3A4 is abundantly expressed in liver and small intestine and is inducible by barbiturates, glucocorticoids and rifampicin.

Specifications

Antigen CYP4Z1
Applications Western Blot
Classification Polyclonal
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene CYP4Z1
Gene Accession No. Q86W10
Gene Alias CYP4A20; CYP4Z1; CYPIVZ1; cytochrome P450 4Z1; cytochrome P450 family 4 subfamily Z member 1; cytochrome P450, family 4, subfamily Z, polypeptide 1; UNQ3060/PRO9882
Gene Symbols CYP4Z1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 89-118 amino acids from the N-terminal region of human CYP4Z1.
Purification Method Ammonium sulfate precipitation, Size-exclusion - Dialysis
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 199974
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less