missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CYP2J2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5106559
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5106559 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5106559 Supplier Invitrogen™ Supplier No. PA5106559
Only null left

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total Cytochrome P450 2J2.

This gene encodes a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This protein localizes to the endoplasmic reticulum and is thought to be the predominant enzyme responsible for epoxidation of endogenous arachidonic acid in cardiac tissue.

Specifications

Antigen CYP2J2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CYP2J2
Gene Accession No. P51589
Gene Alias Albendazole monooxygenase (hydroxylating); Albendazole monooxygenase (sulfoxide-forming); Arachidonic acid epoxygenase; CPJ2; CYP2J2; Cyp2j4; CYPIIJ2; cytochrome P450 2J2; cytochrome P450 family 2 subfamily J member 2; cytochrome P450, family 2, subfamily J, polypeptide 2; cytochrome P450, family 2, subfamily j, polypeptide 4; cytochrome P450, subfamily IIJ (arachidonic acid epoxygenase) polypeptide 2; flavoprotein-linked monooxygenase; Hydroperoxy icosatetraenoate isomerase; microsomal monooxygenase
Gene Symbols CYP2J2
Host Species Rabbit
Immunogen A synthesized peptide derived from human CYP2J2(Accession P51589), corresponding to amino acid residues L240-G290.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1573
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less