missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CYP2C9 Recombinant Rabbit Monoclonal Antibody (23GB5885)
GREENER_CHOICE

Catalog No. MA552938
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA552938 100 μL
MA552939 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA552938 Supplier Invitrogen™ Supplier No. MA552938
Only null left

Rabbit Recombinant Monoclonal Antibody

CYP2C9 Recombinant Monoclonal Antibody for Flow, ICC/IF, Western Blot

CYP2C9 is a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This protein localizes to the endoplasmic reticulum and its expression is induced by rifampin. The enzyme is known to metabolize many xenobiotics, including phenytoin, tolbutamide, ibuprofen and S-warfarin.

Specifications

Antigen CYP2C9
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB5885
Concentration 0.33 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CYP2C9
Gene Accession No. P11712
Gene Alias (R)-limonene 6-monooxygenase; (S)-limonene 6-monooxygenase; (S)-limonene 7-monooxygenase; Cholesterol 25-hydroxylase; CPC9; CYP2C; CYP2C10; CYP2C9; CYPIIC9; Cytochrome P450 2C9; cytochrome P450 family 2 subfamily C member 9; Cytochrome P450 MP-4; Cytochrome P450 MP-8; Cytochrome P450 PB-1; cytochrome P-450 S-mephenytoin 4-hydroxylase; cytochrome P450, family 2, subfamily C, polypeptide 9; cytochrome P-450MP; flavoprotein-linked monooxygenase; MGC149605; MGC88320; microsomal monooxygenase; P450IIC9; S-mephenytoin 4-hydroxylase; xenobiotic monooxygenase
Gene Symbols CYP2C9
Host Species Rabbit
Immunogen A synthesized peptide derived from human Cytochrome P450 2C9 (440-490AA).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1559
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less