missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CYFIP1 Recombinant Rabbit Monoclonal Antibody (23GB1605)

Catalog No. MA553853
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA553853 20 μL
MA553852 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA553853 Supplier Invitrogen™ Supplier No. MA553853
Only null left

Rabbit Recombinant Monoclonal Antibody

CYFIP1 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow

Component of the CYFIP1-EIF4E-FMR1 complex which binds to the mRNA cap and mediates translational repression. In the CYFIP1-EIF4E-FMR1 complex this subunit is an adapter between EIF4E and FMR1. Promotes the translation repression activity of FMR1 in brain probably by mediating its association with EIF4E and mRNA. Involved in formation of membrane ruffles and lamellipodia protrusions and in axon outgrowth. Binds to F-actin but not to RNA. Regulator of epithelial morphogenesis. May act as an invasion suppressor in cancers.

Specifications

Antigen CYFIP1
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB1605
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Cyfip1
Gene Accession No. Q7L576, Q7TMB8
Gene Alias CY; CYFIP; cyfip1; cyfip1 protein; cytoplasmic FMR1 interacting protein 1; cytoplasmic FMR1-interacting protein 1; Cytoplasmic FMR1-interacting protein 1 homolog; cytoplasmic FMRP interacting protein 1; E030028J09Rik; fb15a06; FLJ45151; FMR1-interacting protein 1; KIAA0068; l(7)1Rl; l71Rl; l7Rl1; LOW QUALITY PROTEIN: cytoplasmic FMR1-interacting protein 1; mKIAA0068; P140sra1; p140sra-1; pl-1; selective hybridizing clone; Shyc; Specifically Rac1-associated protein 1; Sra1; Sra-1; Unknown (protein for IMAGE:7152255); wu:fb15a06
Gene Symbols Cyfip1
Host Species Rabbit
Immunogen A synthesized peptide derived from human CYFIP1 (50-100AA).
Purification Method Affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20430, 23191, 308666
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less