missing translation for 'onlineSavingsMsg'
Learn More

Cytiva CyDye™ DIGE Fluors: Cy2

Catalog No. 45002259
Change view
Click to view available options
Quantity:
5 nM
5 x 5 nM
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45002259 5 nM
45002244 5 x 5 nM
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 45002259 Supplier Cytiva Supplier No. 25801082

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Allow detection of up to three prelabeled protein samples and standards on the same 2-D electrophoresis gel

  • Size- and charge-matched dyes enable co-migration of labeled samples within the gel
  • Bright and highly sensitive dyes allow the use of the minimal labeling technique
  • Minimal loss of signal during labeling, separation, and scanning
  • No change in signal over wide pH range used during first-dimension (IEF) separation
  • Discrete signal from each fluor with minimal cross-talk contributes to high accuracy
  • Allow the labeling of 50μg protein per labeling reaction

For general 2-D applications where sufficient amounts of sample are available.

Specifications

Format Solid
For Use With (Application) For general 2-D applications where sufficient amounts of sample are available, detection of up to three prelabeled protein samples and standards on the same 2-D electrophoresis gel
Quantity 5 nM
Product Type Amersham CyDye DIGE Fluor Cy2 Minimal Dye
Kit Contents 5nM of CyDye DIGE Fluor Cy2 minimal dye
Content And Storage -15°C to -30°C
Color Blue