missing translation for 'onlineSavingsMsg'
Learn More

Medisca Cyclosporine A USP

Catalog No. 14185663
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14185663 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14185663 Supplier Medisca Supplier No. 066004
Get 1 Extra Chemical When You Buy 4
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Cyclosporine A USP is a white to almost white powder. Alternative name(s): ciclosporin, cyclosporin, Cyclosporin A

Cyclosporine A USP is a white to almost white powder.

Alternative name(s): ciclosporin, cyclosporin, Cyclosporin A

Specifications

Chemical Name or Material Cyclosporine A USP
Packaging Jar 100ml, White, HDPE, Round, 38-400, TRIVENI-DB, CT
CAS 59865-13-3
Type Drug Standard
Molecular Weight (g/mol) 1202.61
Quantity 25 g
Percent Purity 97 to 101.5 %
Grade USP