missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Medisca Cyclosporine A USP
Description
Cyclosporine A USP is a white to almost white powder.
Alternative name(s): ciclosporin, cyclosporin, Cyclosporin A
Specifications
Specifications
| Chemical Name or Material | Cyclosporine A USP |
| Packaging | Jar 1500ml, White, HDPE, Round, 89-400, TRIVENI-DB, CT |
| CAS | 59865-13-3 |
| Type | Drug Standard |
| Molecular Weight (g/mol) | 1202.61 |
| Quantity | 500 g |
| Percent Purity | 97 to 101.5 % |
| Grade | USP |