missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Cyclosporin A, 98%

Cyclosporin A, 98%, CAS # 59865-13-3, is a compound that shows immunosuppressive, antifungal, antirheumatic, and dermatologic activities. It is also a calcineurin and enzyme inhibitor. | CAS: 59865-13-3 | C62H111N11O12 | 1202.64 g/mol

$391.36

Chemical Identifiers

CAS 59865-13-3
Molecular Formula C62H111N11O12
Molecular Weight (g/mol) 1202.64
MDL Number MFCD00274558
InChI Key PMATZTZNYRCHOR-IMVLJIQENA-N
PubChem CID 132274082
SMILES CCC1NC(=O)C(C(O)C(C)C\C=C\C)N(C)C(=O)C(C(C)C)N(C)C(=O)C(CC(C)C)N(C)C(=O)C(CC(C)C)N(C)C(=O)C(C)NC(=O)C(C)NC(=O)C(CC(C)C)N(C)C(=O)C(NC(=O)C(CC(C)C)N(C)C(=O)CN(C)C1=O)C(C)C
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AC457970010
View Documents Promotion Details
Thermo Scientific Chemicals
457970010
1 g
Each for $391.36
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Cyclosporin A is a non-polar cyclic oligopeptide produced by the fungus Tolypocladium inflatum

Specifications

Chemical Identifiers

59865-13-3
1202.64
PMATZTZNYRCHOR-IMVLJIQENA-N
CCC1NC(=O)C(C(O)C(C)C\C=C\C)N(C)C(=O)C(C(C)C)N(C)C(=O)C(CC(C)C)N(C)C(=O)C(CC(C)C)N(C)C(=O)C(C)NC(=O)C(C)NC(=O)C(CC(C)C)N(C)C(=O)C(NC(=O)C(CC(C)C)N(C)C(=O)CN(C)C1=O)C(C)C
C62H111N11O12
MFCD00274558
132274082

Specifications

Cyclosporin A
146.0°C to 151.0°C
2% max. (60°C, 3 hrs, 5 mm Hg)
MFCD00274558
CCC1NC(=O)C(C(O)C(C)C\C=C\C)N(C)C(=O)C(C(C)C)N(C)C(=O)C(CC(C)C)N(C)C(=O)C(CC(C)C)N(C)C(=O)C(C)NC(=O)C(C)NC(=O)C(CC(C)C)N(C)C(=O)C(NC(=O)C(CC(C)C)N(C)C(=O)CN(C)C1=O)C(C)C
1202.64
1202.61
Cyclosporin A
59865-13-3
Authentic
C62H111N11O12
PMATZTZNYRCHOR-IMVLJIQENA-N
30-ethyl-33-[(4E)-1-hydroxy-2-methylhex-4-en-1-yl]-1,4,7,10,12,15,19,25,28-nonamethyl-6,9,18,24-tetrakis(2-methylpropyl)-3,21-bis(propan-2-yl)-1,4,7,10,13,16,19,22,25,28,31-undecaazacyclotritriacontan-2,5,8,11,14,17,20,23,26,29,32-undecone
132274082
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement Harmful if swallowed. May cause cancer. May damage fertility. May damage the unborn child.

GHS P Statement Wash face, hands and any exposed skin thoroughly after handling. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Obtain special instructions before use. Use personal protective equipment as required. IF exposed or concerned: Get medical advice/attention.

Danger

SDS
Documents

Documents

RUO – Research Use Only