missing translation for 'onlineSavingsMsg'
Learn More

Cyclopropyltriphenylphosphonium Bromide 97.0+%, TCI America™

Catalog No. C13785G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C13785G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C13785G Supplier TCI America Supplier No. C13785G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 14114-05-7
Molecular Formula C21H20BrP
Molecular Weight (g/mol) 383.27
MDL Number MFCD00011872
InChI Key XMPWFKHMCNRJCL-UHFFFAOYSA-M
Synonym cyclopropyltriphenylphosphonium bromide, phosphonium, cyclopropyltriphenyl-, bromide, cyclopropyltriphenylphosphanium bromide, cyclopropyl triphenyl phosphanium bromide, acmc-209cls, cyclopropyltriphenylphosphoniumbromide, cyclopropyl triphenylphosphonium bromide, cyclopropyl-triphenylphosphonium bromide, cyclopropyltriphenyl-phosphonium bromide, triphenyl-cyclopropylphosphonium bromide
PubChem CID 2723931
IUPAC Name cyclopropyltriphenylphosphanium bromide
SMILES [Br-].C1CC1[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 185°C
Color White-Yellow
Quantity 5 g
Formula Weight 383.27
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Cyclopropyltriphenylphosphonium Bromide