missing translation for 'onlineSavingsMsg'
Learn More

Cyclopropyltriphenylphosphonium bromide, 98%

Catalog No. AAB2510222
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2510222 100 g
AAB2510206 5 g
AAB2510214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2510222 Supplier Thermo Scientific Chemicals Supplier No. B2510222
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 14114-05-7 | C21H20BrP | 383.27 g/mol

It is used for electrolytic polarization of Zn, has potential antitumor activity, reactant for synthesis of cycloporpylidenemethylarylethynynes, keto halides for sequential azidation and intramolecular Schmidt reactions, a reactant for hydroalkynylation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used for electrolytic polarization of Zn, has potential antitumor activity, reactant for synthesis of cycloporpylidenemethylarylethynynes, keto halides for sequential azidation and intramolecular Schmidt reactions, a reactant for hydroalkynylation.

Solubility
Soluble in water.

Notes
Hygroscopic. Store under inert gas. Store away from oxidizing agents, water/ moisture.

Chemical Identifiers

CAS 14114-05-7
Molecular Formula C21H20BrP
Molecular Weight (g/mol) 383.27
MDL Number MFCD00011872
InChI Key XMPWFKHMCNRJCL-UHFFFAOYSA-M
Synonym cyclopropyltriphenylphosphonium bromide, phosphonium, cyclopropyltriphenyl-, bromide, cyclopropyltriphenylphosphanium bromide, cyclopropyl triphenyl phosphanium bromide, acmc-209cls, cyclopropyltriphenylphosphoniumbromide, cyclopropyl triphenylphosphonium bromide, cyclopropyl-triphenylphosphonium bromide, cyclopropyltriphenyl-phosphonium bromide, triphenyl-cyclopropylphosphonium bromide
PubChem CID 2723931
IUPAC Name cyclopropyl(triphenyl)phosphanium;bromide
SMILES [Br-].C1CC1[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 183°C to 186°C
Quantity 100 g
Beilstein 6413483
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 383.27
Percent Purity 98%
Chemical Name or Material Cyclopropyltriphenylphosphonium bromide
Hazard Category H312+H332
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P271-P280-P302+P352-P304+P340-P312-P363-P501c
EINECSNumber 237-970-1
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only