missing translation for 'onlineSavingsMsg'
Learn More

(Cyclopropylmethyl)triphenylphosphonium bromide, 98+%

Catalog No. p-7043527
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0114506 5 g
AAL0114514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0114506 Supplier Thermo Scientific Chemicals Supplier No. L0114506

CAS: 14799-82-7 | C22H22BrP | 397.30 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 14799-82-7
Molecular Formula C22H22BrP
Molecular Weight (g/mol) 397.30
MDL Number MFCD00051878
InChI Key WFQSHRSBITUSIB-UHFFFAOYSA-M
Synonym cyclopropylmethyl triphenylphosphonium bromide, cyclopropylmethyl triphenyl phosphanium bromide, cyclopropylmethyl triphenyl phosphonium bromide, phosphonium, cyclopropylmethyl triphenyl-, bromide, cyclopropylmethyl triphenylphosphanium bromide, acmc-1cg9v, cyclopropylmethyltriphenylphosphonium bromide, cyclopropylmethyl triphenylphosphoniumbromide, phosphonium, cyclopropylmethyl triphenyl-, bromide 1:1
PubChem CID 3084208
IUPAC Name cyclopropylmethyl(triphenyl)phosphanium;bromide
SMILES [Br-].C(C1CC1)[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 178°C to 182°C
Quantity 5 g
Beilstein 4068302
Sensitivity Hygroscopic
Formula Weight 397.29
Percent Purity ≥98%
Chemical Name or Material (Cyclopropylmethyl)triphenylphosphonium bromide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 238-862-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only