missing translation for 'onlineSavingsMsg'
Learn More

Cyclopropylacetylene 98.0+%, TCI America™

Catalog No. p-7125221
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C19845G 5 g
C198425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C19845G Supplier TCI America Supplier No. C19845G
Only null left

Chemical Identifiers

CAS 6746-94-7
Molecular Formula C5H6
Molecular Weight (g/mol) 66.103
MDL Number MFCD02181090
InChI Key NPTDXPDGUHAFKC-UHFFFAOYSA-N
Synonym cyclopropylacetylene, cyclopropyl acetylene, ethynyl cyclopropane, cyclopropane,ethynyl, cyclopropane, ethynyl, cyclopropylethyne, cyclopropyacetylene, ethynyl-cyclopropane, ethynylcyclo-propane, cyclopropylacethylene
PubChem CID 138823
IUPAC Name ethynylcyclopropane
SMILES C#CC1CC1

Specifications

Color Colorless
Boiling Point 52°C
Quantity 5 g
UN Number 1993
Formula Weight 66.10
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Cyclopropylacetylene
EINECSNumber (3)-4334
TSCA No