accessibility menu, dialog, popup

UserName

CH

CAS RN 6746-94-7

IUPAC Name: ethynylcyclopropane
Synonyms: ethynylcyclopropane
Molecular Weight (g/mol): 66.1
Molecular Formula: C5H6
InChi Key: NPTDXPDGUHAFKC-UHFFFAOYSA-N
SMILES: C#CC1CC1
Boiling Point 
  • (3)
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (3)
  • (5)
Percent Purity 
  • (2)
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (6)
  • (3)
  • (3)
  • (2)
  • (3)
  • (5)
  • (2)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (5)