missing translation for 'onlineSavingsMsg'
Learn More

Cyclopropylacetylene, 97%

Catalog No. p-7035714
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5344006 5 g
AAH5344014 25 g
AAH5344022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH5344006 Supplier Thermo Scientific Chemicals Supplier No. H5344006
Only null left

CAS: 6746-94-7 | C5H6 | 66.103 g/mol

Cyclopropylacetylene is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclopropylacetylene is used as an organic chemical synthesis intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.

Chemical Identifiers

CAS 6746-94-7
Molecular Formula C5H6
Molecular Weight (g/mol) 66.103
MDL Number MFCD02181090
InChI Key NPTDXPDGUHAFKC-UHFFFAOYSA-N
Synonym cyclopropylacetylene, cyclopropyl acetylene, ethynyl cyclopropane, cyclopropane,ethynyl, cyclopropane, ethynyl, cyclopropylethyne, cyclopropyacetylene, ethynyl-cyclopropane, ethynylcyclo-propane, cyclopropylacethylene
PubChem CID 138823
IUPAC Name ethynylcyclopropane
SMILES C#CC1CC1

Specifications

Density 0.781
Boiling Point 51°C
Flash Point −17°C (−1°F)
Refractive Index 1.429
Quantity 5 g
UN Number UN3295
Solubility Information Slightly soluble in water.
Formula Weight 66.1
Percent Purity 97%
Chemical Name or Material Cyclopropylacetylene
Hazard Category H225-H315-H318
Hazard Statement GHS H Statement
H225-H318-H315
Highly flammable liquid and vapor.
Causes serious eye damage.
Causes skin irritation.
Precautionary Statement P210-P233-P240-P241-P242-P243-P264b-P280-P303+P361+P353-P305+P351+P338-P310-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only