missing translation for 'onlineSavingsMsg'
Learn More

Cyclopropylacetic acid, 97%

Catalog No. AC443780050
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC443780050 5 g
AC443780010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC443780050 Supplier Thermo Scientific Chemicals Supplier No. 443780050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 5239-82-7 | C5H8O22 | 100.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5239-82-7
Molecular Formula C5H8O22
Molecular Weight (g/mol) 100.12
MDL Number MFCD00041544
InChI Key BXXOHRWKRXBZTG-UHFFFAOYSA-N
Synonym cyclopropylacetic acid, cyclopropaneacetic acid, cyclopropyl acetic acid, cyclopropyl-acetic acid, carboxymethyl cyclopropane, 2-cyclopropyl-acetic acid, 2-cyclopropylethanoic acid, acetic cyclopropyl acid, pubchem19703, 2-cyclopropyl acetic acid
PubChem CID 138440
IUPAC Name 2-cyclopropylacetic acid
SMILES CN1CCN(C)B1N=S=NB1N(C)CCN1C

Specifications

Density 1.1400g/mL
Boiling Point 60.0°C to 61.0°C (29.0 mmHg)
Flash Point 93°C
Assay Percent Range 97%
Quantity 5 g
Specific Gravity 1.14
Formula Weight 100.12
Percent Purity 97%
Chemical Name or Material Cyclopropylacetic acid
Hazard Category GHS Signal Word: Danger

RUO – Research Use Only