accessibility menu, dialog, popup

UserName

CAS RN 5239-82-7

IUPAC Name: 2-cyclopropylacetic acid
Synonyms: —
Molecular Weight (g/mol): 100.12
Molecular Formula: C5H8O22
InChi Key: BXXOHRWKRXBZTG-UHFFFAOYSA-N
SMILES: CN1CCN(C)B1N=S=NB1N(C)CCN1C
Boiling Point 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results