missing translation for 'onlineSavingsMsg'
Learn More

Cyclopropylacetic acid, 97%

Catalog No. AAL0941603
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0941603 1 g
AAL0941606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0941603 Supplier Thermo Scientific Chemicals Supplier No. L0941603
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 5239-82-7 | C5H8O22 | 100.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5239-82-7
Molecular Formula C5H8O22
Molecular Weight (g/mol) 100.12
MDL Number MFCD00041544
InChI Key BXXOHRWKRXBZTG-UHFFFAOYSA-N
Synonym cyclopropylacetic acid, cyclopropaneacetic acid, cyclopropyl acetic acid, cyclopropyl-acetic acid, carboxymethyl cyclopropane, 2-cyclopropyl-acetic acid, 2-cyclopropylethanoic acid, acetic cyclopropyl acid, pubchem19703, 2-cyclopropyl acetic acid
PubChem CID 138440
IUPAC Name 2-cyclopropylacetic acid
SMILES CN1CCN(C)B1N=S=NB1N(C)CCN1C

Specifications

Density 1.076
Boiling Point 189°C to 191°C
Flash Point 93°C (199°F)
Odor Pungent
Refractive Index 1.435
Quantity 1 g
UN Number UN3265
Beilstein 2323311
Formula Weight 100.12
Percent Purity 97%
Chemical Name or Material Cyclopropylacetic acid
Show More Show Less
Hazard Category H227-H314-H335
Hazard Statement GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Precautionary Statement P210-P235-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only