missing translation for 'onlineSavingsMsg'
Learn More

Cyclopropyl Phenyl Sulfide 98.0+%, TCI America™

Catalog No. p-7124743
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C10145G 5 g
C101425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C10145G Supplier TCI America Supplier No. C10145G
Only null left

Chemical Identifiers

CAS 14633-54-6
Molecular Formula C9H10S
Molecular Weight (g/mol) 150.24
MDL Number MFCD00009684
InChI Key YIBKCPJOFAUAKY-UHFFFAOYSA-N
Synonym cyclopropyl phenyl sulfide, cyclopropyl phenyl sulfane, cyclopropylsulfanyl benzene, benzene, cyclopropylthio, cyclopropylphenylsulfide, cyclopropyl phenyl sulphide, unii-72c99lp48p, cyclopropylsulfanyl-benzene, pubchem10736, cyclopropylphenyl sulfide
PubChem CID 123348
IUPAC Name (cyclopropylsulfanyl)benzene
SMILES C1CC1SC1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 90°C
Quantity 5 g
Formula Weight 150.24
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Cyclopropyl Phenyl Sulfide