missing translation for 'onlineSavingsMsg'
Learn More

Cyclopropyl phenyl sulfide, 98+%

Catalog No. AAL1232106
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1232106 5 g
AAL1232103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1232106 Supplier Thermo Scientific Chemicals Supplier No. L1232106
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 14633-54-6 | C9H10S | 150.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 14633-54-6
Molecular Formula C9H10S
Molecular Weight (g/mol) 150.24
MDL Number MFCD00009684
InChI Key YIBKCPJOFAUAKY-UHFFFAOYSA-N
Synonym cyclopropyl phenyl sulfide, cyclopropyl phenyl sulfane, cyclopropylsulfanyl benzene, benzene, cyclopropylthio, cyclopropylphenylsulfide, cyclopropyl phenyl sulphide, unii-72c99lp48p, cyclopropylsulfanyl-benzene, pubchem10736, cyclopropylphenyl sulfide
PubChem CID 123348
IUPAC Name cyclopropylsulfanylbenzene
SMILES C1CC1SC1=CC=CC=C1

Specifications

Density 1.045
Boiling Point 61°C to 62°C (1 mmHg)
Flash Point 91°C (195°F)
Odor Stench
Refractive Index 1.581
Quantity 5 g
UN Number UN3334
Beilstein 2075701
Formula Weight 150.24
Percent Purity ≥98%
Chemical Name or Material Cyclopropyl phenyl sulfide
Show More Show Less
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H227
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: AVIATION REGULATED LIQUID, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only