missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclopropyl Methyl Ketone 98.0+%, TCI America™
Supplier: TCI America C0985500ML
Specifications
| Cyclopropyl Methyl Ketone | |
| Yellow | |
| C5H8O | |
| 500 mL | |
| cyclopropyl methyl ketone, acetylcyclopropane, methyl cyclopropyl ketone, ethanone, 1-cyclopropyl, 1-cyclopropyl-ethanone, 1-cyclopropylethan-1-one, ketone, cyclopropyl methyl, cyclopropylmethylketone, unii-n27yy1xcfh, cyclopropylethanone | |
| CC(=O)C1CC1 | |
| 84.118 | |
| 84.12 | |
| Liquid |
| 765-43-5 | |
| 110°C | |
| MFCD00001297 | |
| 1224 | |
| HVCFCNAITDHQFX-UHFFFAOYSA-N | |
| 1-cyclopropylethanone | |
| 13004 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 765-43-5 | |
| 84.118 | |
| HVCFCNAITDHQFX-UHFFFAOYSA-N | |
| 13004 | |
| CC(=O)C1CC1 |
| C5H8O | |
| MFCD00001297 | |
| cyclopropyl methyl ketone, acetylcyclopropane, methyl cyclopropyl ketone, ethanone, 1-cyclopropyl, 1-cyclopropyl-ethanone, 1-cyclopropylethan-1-one, ketone, cyclopropyl methyl, cyclopropylmethylketone, unii-n27yy1xcfh, cyclopropylethanone | |
| 1-cyclopropylethanone |
Safety and Handling
TSCA : No