accessibility menu, dialog, popup

UserName

CAS RN 765-43-5

IUPAC Name: 1-cyclopropylethan-1-one
Synonyms: cyclopropyl methyl ketone
Molecular Weight (g/mol): 84.12
Molecular Formula: C5H8O
InChi Key: HVCFCNAITDHQFX-UHFFFAOYSA-N
SMILES: CC(=O)C1CC1
Boiling Point 
  • (3)
  • (3)
  • (3)
Molecular Weight (g/mol) 
  • (6)
  • (3)
Percent Purity 
  • (3)
  • (3)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (3)
  • (6)
  • (3)
  • (3)
  • (3)
  • (6)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (6)