missing translation for 'onlineSavingsMsg'
Learn More

Cyclopropyl Methyl Ketone 98.0+%, TCI America™

Catalog No. C098525ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C098525ML 25 mL
C0985100ML 100 mL
C0985500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C098525ML Supplier TCI America Supplier No. C098525ML

Chemical Identifiers

CAS 765-43-5
Molecular Formula C5H8O
Molecular Weight (g/mol) 84.118
MDL Number MFCD00001297
InChI Key HVCFCNAITDHQFX-UHFFFAOYSA-N
Synonym cyclopropyl methyl ketone, acetylcyclopropane, methyl cyclopropyl ketone, ethanone, 1-cyclopropyl, 1-cyclopropyl-ethanone, 1-cyclopropylethan-1-one, ketone, cyclopropyl methyl, cyclopropylmethylketone, unii-n27yy1xcfh, cyclopropylethanone
PubChem CID 13004
IUPAC Name 1-cyclopropylethanone
SMILES CC(=O)C1CC1

Specifications

Color Yellow
Boiling Point 110°C
Quantity 25 mL
UN Number 1224
Formula Weight 84.12
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Cyclopropyl Methyl Ketone