missing translation for 'onlineSavingsMsg'
Learn More

Cyclopropyl methyl ketone, 98%

Catalog No. AC111710250
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC111710250 25 mL Glass bottle
AC111711000 100 mL Glass bottle
AC111715000 500 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC111710250 Supplier Thermo Scientific Chemicals Supplier No. 111710250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 765-43-5 | C5H8O | 84.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 765-43-5
Molecular Formula C5H8O
Molecular Weight (g/mol) 84.12
MDL Number MFCD00001297
InChI Key HVCFCNAITDHQFX-UHFFFAOYSA-N
Synonym cyclopropyl methyl ketone, acetylcyclopropane, methyl cyclopropyl ketone, ethanone, 1-cyclopropyl, 1-cyclopropyl-ethanone, 1-cyclopropylethan-1-one, ketone, cyclopropyl methyl, cyclopropylmethylketone, unii-n27yy1xcfh, cyclopropylethanone
PubChem CID 13004
IUPAC Name 1-cyclopropylethanone
SMILES CC(=O)C1CC1

Specifications

Melting Point -68.0°C
Color Colorless to Yellow
Density 0.8980g/mL
Boiling Point 111.0°C
Flash Point 21°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4220 to 1.4260
Linear Formula C3H5COCH3
Quantity 25 mL
Beilstein 07,7
Fieser 01,676
Specific Gravity 0.898
Solubility Information Solubility in water: 185g/L (20°C). Other solubilities: soluble in alcohols,chloride solvents
Viscosity 0.65 mPa.s (20°C)
Formula Weight 84.12
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Cyclopropyl methyl ketone
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Wear protective g
EINECSNumber 212-146-4

RUO – Research Use Only