missing translation for 'onlineSavingsMsg'
Learn More

Cyclopropyl isothiocyanate, 97%

Catalog No. AAL1198503
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1198503 1 g
AAL1198506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1198503 Supplier Thermo Scientific Chemicals Supplier No. L1198503
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 56601-42-4 | C4H5NS | 99.15 g/mol

Cyclopropyl isothiocyanate is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclopropyl isothiocyanate is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to moisture.

Chemical Identifiers

CAS 56601-42-4
Molecular Formula C4H5NS
Molecular Weight (g/mol) 99.15
MDL Number MFCD00040874
InChI Key JGFBQFKZKSSODQ-UHFFFAOYSA-N
Synonym cyclopropyl isothiocyanate, cyclopropane, isothiocyanato, isothiocyanato-cyclopropane, cyclopropylisothiocyanate, acmc-20albt, pubchem22785, cyclopropyl iso-thiocyanate, cyclopropane,isothiocyanato
PubChem CID 92463
IUPAC Name isothiocyanatocyclopropane
SMILES S=C=NC1CC1

Specifications

Density 1.07
Boiling Point 87°C to 88°C (35 mmHg)
Flash Point 57°C (134°F)
Refractive Index 1.548
Quantity 1 g
UN Number UN2929
Beilstein 3600974
Sensitivity Moisture sensitive
Formula Weight 99.16
Percent Purity 97%
Chemical Name or Material Cyclopropyl isothiocyanate
Show More Show Less
Hazard Category H226-H301+H331-H314
Hazard Statement GHS H Statement
H301-H330-H314-H318-H226
Toxic if swallowed.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Hazard Class: 3; Packaging Group: II
EINECSNumber 260-278-6
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only