missing translation for 'onlineSavingsMsg'
Learn More

Cyclopropyl Cyanide 97.0+%, TCI America™

Catalog No. p-7124732
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C09975ML 5 mL
C099725ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C09975ML Supplier TCI America Supplier No. C09975ML
Only null left

Chemical Identifiers

CAS 5500-21-0
Molecular Formula C4H5N
Molecular Weight (g/mol) 67.091
MDL Number MFCD00001269
InChI Key AUQDITHEDVOTCU-UHFFFAOYSA-N
Synonym cyclopropyl cyanide, cyanocyclopropane, cyclopropylnitrile, cyclopropanenitrile, cyclopropane cyanide, 1-cyanocyclopropane, cyclopropane carbonitrile, cyclopropylcyanide, 1-cyanocyclopropyl, cyclopropancarbonitril
PubChem CID 79637
IUPAC Name cyclopropanecarbonitrile
SMILES C1CC1C#N

Specifications

Boiling Point 135°C
Quantity 5 mL
UN Number 3275
Formula Weight 67.09
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Cyclopropyl Cyanide