missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclopropyl 2-Fluorobenzyl Ketone 98.0+%, TCI America™
Supplier: TCI America C271025G
Specifications
| Cyclopropyl 2-Fluorobenzyl Ketone | |
| Yellow | |
| C11H11FO | |
| 25 g | |
| C1CC1C(=O)CC2=CC=CC=C2F | |
| 178.206 | |
| 178.21 | |
| Liquid |
| 150322-73-9 | |
| 61°C | |
| MFCD10687164 | |
| DWBGTJUQWKWYGB-UHFFFAOYSA-N | |
| 1-cyclopropyl-2-(2-fluorophenyl)ethanone | |
| 15289941 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 150322-73-9 | |
| 178.206 | |
| DWBGTJUQWKWYGB-UHFFFAOYSA-N | |
| 1-cyclopropyl-2-(2-fluorophenyl)ethanone |
| C11H11FO | |
| MFCD10687164 | |
| 15289941 | |
| C1CC1C(=O)CC2=CC=CC=C2F |
Safety and Handling
TSCA : No