missing translation for 'onlineSavingsMsg'
Learn More

Cyclopropanethiocarboxamide, 97%

Catalog No. AAH3231503
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3231503 1 g
AAH3231506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3231503 Supplier Thermo Scientific Chemicals Supplier No. H3231503
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 20295-34-5 | C4H7NS | 101.167 g/mol

Cyclopropanecarbothioamide is used for purification in single-step preparation of thiazolo [5, 4-b] pyridine-and thiazolo [5, 4-c] pyridine derivatives from chloronitropyridines and thioamides, or thioureas.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclopropanecarbothioamide is used for purification in single-step preparation of thiazolo [5, 4-b] pyridine-and thiazolo [5, 4-c] pyridine derivatives from chloronitropyridines and thioamides, or thioureas.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 20295-34-5
Molecular Formula C4H7NS
Molecular Weight (g/mol) 101.167
MDL Number MFCD09469287
InChI Key IIPJWNFOLPDTEQ-UHFFFAOYSA-N
Synonym cyclopropanethiocarboxamide, cyclopropanecarbothioic acid amide, cyclopropanethioamide, cyclopropane carbothioamide, cyclopropane-thiocarboxamide, cyclopropane-1-carbothioamide, aminocyclopropylmethane-1-thione
PubChem CID 22140884
IUPAC Name cyclopropanecarbothioamide
SMILES C1CC1C(=S)N

Specifications

Melting Point 111°C to 113°C
Quantity 1 g
Formula Weight 101.17
Percent Purity 97%
Chemical Name or Material Cyclopropanethiocarboxamide
Hazard Category H302
Hazard Statement GHS H Statement
H301
Toxic if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
Recommended Storage Ambient temperatures

RUO – Research Use Only