missing translation for 'onlineSavingsMsg'
Learn More

Cyclopropanesulfonyl Chloride 97.0+%, TCI America™

Catalog No. c21871g
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
Catalog No. Quantity
C21871G 1 g
C21875G 5 g
2 options

Catalog No. C21871G

Supplier: TCI America C21871G

Only null left

Chemical Identifiers

CAS 139631-62-2
Molecular Formula C3H5ClO2S
Molecular Weight (g/mol) 140.58
MDL Number MFCD01631933
InChI Key PFWWSGFPICCWGU-UHFFFAOYSA-N
Synonym cyclopropanesulfonylchloride, chlorocyclopropylsulfone, cyclopropanesulphonyl chloride, cyclopropylsulfonyl chloride, cyclopropylsulphonyl chloride, cyclopropane sulfonyl chloride, cyclopropanesulfonic acid chloride, pubchem10869, acmc-209cje, cyclopropylsulfonylchloride
PubChem CID 2760919
IUPAC Name cyclopropanesulfonyl chloride
SMILES ClS(=O)(=O)C1CC1

Specifications

Color Yellow
Boiling Point 60°C
Quantity 1 g
UN Number 3265
Formula Weight 140.58
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Cyclopropanesulfonyl Chloride
TSCA No
Recommended Storage Refrigerator