accessibility menu, dialog, popup

UserName

CAS RN 139631-62-2

IUPAC Name: cyclopropanesulfonyl chloride
Synonyms: cyclopropanesulfonyl chloride
Molecular Weight (g/mol): 140.58
Molecular Formula: C3H5ClO2S
InChi Key: PFWWSGFPICCWGU-UHFFFAOYSA-N
SMILES: ClS(=O)(=O)C1CC1
Boiling Point 
  • (2)
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (3)
  • (3)
  • (4)