missing translation for 'onlineSavingsMsg'
Learn More

Cyclopropanesulfonyl chloride, 97%

Catalog No. AAH2910706
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2910706 5 g
AAH2910703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2910706 Supplier Thermo Scientific Chemicals Supplier No. H2910706
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 139631-62-2 | C3H5ClO2S | 140.58 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 139631-62-2
Molecular Formula C3H5ClO2S
Molecular Weight (g/mol) 140.58
MDL Number MFCD01631933
InChI Key PFWWSGFPICCWGU-UHFFFAOYSA-N
Synonym cyclopropanesulfonylchloride, chlorocyclopropylsulfone, cyclopropanesulphonyl chloride, cyclopropylsulfonyl chloride, cyclopropylsulphonyl chloride, cyclopropane sulfonyl chloride, cyclopropanesulfonic acid chloride, pubchem10869, acmc-209cje, cyclopropylsulfonylchloride
PubChem CID 2760919
IUPAC Name cyclopropanesulfonyl chloride
SMILES ClS(=O)(=O)C1CC1

Specifications

Density 1.39
Boiling Point 60°C (2 mmHg)
Flash Point 96°C (205°F)
Refractive Index 1.477
Quantity 5 g
UN Number UN3265
Sensitivity Moisture sensitive
Formula Weight 140.59
Percent Purity 97%
Chemical Name or Material Cyclopropanesulfonyl chloride
Hazard Category H302+H332-H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only