missing translation for 'onlineSavingsMsg'
Learn More

Cyclopropanesulfonyl chloride, 97%, Thermo Scientific™

Chemical Identifiers

CAS 139631-62-2
Molecular Formula C3H5ClO2S
Molecular Weight (g/mol) 140.58
MDL Number MFCD01631933
InChI Key PFWWSGFPICCWGU-UHFFFAOYSA-N
Synonym cyclopropanesulfonylchloride, chlorocyclopropylsulfone, cyclopropanesulphonyl chloride, cyclopropylsulfonyl chloride, cyclopropylsulphonyl chloride, cyclopropane sulfonyl chloride, cyclopropanesulfonic acid chloride, pubchem10869, acmc-209cje, cyclopropylsulfonylchloride
PubChem CID 2760919
IUPAC Name cyclopropanesulfonyl chloride
SMILES ClS(=O)(=O)C1CC1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC439020010
SDS View Documents
Thermo Scientific Chemicals
439020010
1g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC439020050
SDS View Documents
Thermo Scientific Chemicals
439020050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

139631-62-2
140.58
PFWWSGFPICCWGU-UHFFFAOYSA-N
2760919
ClS(=O)(=O)C1CC1
C3H5ClO2S
MFCD01631933
cyclopropanesulfonylchloride, chlorocyclopropylsulfone, cyclopropanesulphonyl chloride, cyclopropylsulfonyl chloride, cyclopropylsulphonyl chloride, cyclopropane sulfonyl chloride, cyclopropanesulfonic acid chloride, pubchem10869, acmc-209cje, cyclopropylsulfonylchloride
cyclopropanesulfonyl chloride

Specifications

139631-62-2
60.0°C (2.0 Torr)
Authentic
Glass bottle
MFCD01631933
1.38
PFWWSGFPICCWGU-UHFFFAOYSA-N
cyclopropanesulfonyl chloride
2760919
97%
1.3800g/mL
96°C
96% min. (GC)
C3H5ClO2S
1g
cyclopropanesulfonylchloride, chlorocyclopropylsulfone, cyclopropanesulphonyl chloride, cyclopropylsulfonyl chloride, cyclopropylsulphonyl chloride, cyclopropane sulfonyl chloride, cyclopropanesulfonic acid chloride, pubchem10869, acmc-209cje, cyclopropylsulfonylchloride
ClS(=O)(=O)C1CC1
140.58
140.59
Cyclopropanesulfonyl chloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Harmful if inhaled.
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER

GHS Signal Word: Danger

SDS
Documents

Documents

RUO – Research Use Only