missing translation for 'onlineSavingsMsg'
Learn More

Cyclopropanemethanol 98.0+%, TCI America™

Catalog No. p-7124722
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C09825ML 5 mL
C098225ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C09825ML Supplier TCI America Supplier No. C09825ML
Only null left

Chemical Identifiers

CAS 2516-33-8
Molecular Formula C4H8O
Molecular Weight (g/mol) 72.11
MDL Number MFCD00001309
InChI Key GUDMZGLFZNLYEY-UHFFFAOYSA-N
Synonym cyclopropanemethanol, cyclopropyl carbinol, cyclopropylcarbinol, cyclopropylmethyl alcohol, hydroxymethyl cyclopropane, cyclopropanemethyl alcohol, cyclopropylcarbinyl alcohol, hydroxymethylcyclopropane, cyclopropyl-methanol, cyclopropane methanol
PubChem CID 75644
IUPAC Name cyclopropylmethanol
SMILES OCC1CC1

Specifications

Color Yellow
Boiling Point 123°C
Quantity 5 mL
UN Number 1987
Formula Weight 72.11
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Cyclopropanemethanol
RTECSNumber GZ2212250
TSCA Yes