accessibility menu, dialog, popup

UserName

CAS RN 2516-33-8

IUPAC Name: cyclopropylmethanol
Synonyms: cyclopropylmethanol
Molecular Weight (g/mol): 72.11
Molecular Formula: C4H8O
InChi Key: GUDMZGLFZNLYEY-UHFFFAOYSA-N
SMILES: OCC1CC1
Boiling Point 
  • (2)
  • (3)
  • (4)
Percent Purity 
  • (2)
  • (3)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (2)
  • (7)
  • (2)
  • (3)
  • (4)
  • (2)
  • (3)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (6)