missing translation for 'onlineSavingsMsg'
Learn More

Cyclopropanemethanol, 98%

Catalog No. AAA1256706
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1256706 5 g
AAA1256714 25 g
AAA1256722 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1256706 Supplier Thermo Scientific Chemicals Supplier No. A1256706
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2516-33-8 | C4H8O | 72.11 g/mol

Cyclopropanemethanol is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclopropanemethanol is used as an organic chemical synthesis intermediate.

Solubility
Fully miscible with water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and acid chlorides.

Chemical Identifiers

CAS 2516-33-8
Molecular Formula C4H8O
Molecular Weight (g/mol) 72.11
MDL Number MFCD00001309
InChI Key GUDMZGLFZNLYEY-UHFFFAOYSA-N
Synonym cyclopropanemethanol, cyclopropyl carbinol, cyclopropylcarbinol, cyclopropylmethyl alcohol, hydroxymethyl cyclopropane, cyclopropanemethyl alcohol, cyclopropylcarbinyl alcohol, hydroxymethylcyclopropane, cyclopropyl-methanol, cyclopropane methanol
PubChem CID 75644
IUPAC Name cyclopropylmethanol
SMILES OCC1CC1

Specifications

Melting Point <-60°C
Density 0.904
Boiling Point 123°C to 124°C
Flash Point 35°C (95°F)
Odor Mild
Refractive Index 1.433
Quantity 5 g
UN Number UN1987
Beilstein 1846846
Solubility Information Fully miscible with water.
Formula Weight 72.11
Percent Purity 98%
Chemical Name or Material Cyclopropanemethanol
Show More Show Less
Hazard Category H226-H302-H314-H335
Hazard Statement GHS H Statement
H301-H311-H226-H373-H332-H319
Toxic if swallowed.
Toxic in contact with skin.
Flammable liquid and vapour.
May cause damage to organs through prolonged or repeated exposure.
Harmful if inhaled.
Causes serious eye irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 219-735-5
RTECSNumber GZ2212250
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only