missing translation for 'onlineSavingsMsg'
Learn More

Cyclopropanecarboxaldehyde, 98%

Catalog No. AAL1493103
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1493103 1 g
AAL1493106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1493103 Supplier Thermo Scientific Chemicals Supplier No. L1493103
Get Up to $1,800 in Chemicals with Purchase
View all promotions

missing translation for 'validMainframeMsg'

CAS: 1489-69-6 | C4H6O | 70.091 g/mol

Cyclopropanecarboxaldehyde reacts with Grignard or organolithium reagents to give the expected secondary alcohols. These are susceptible to ring opening in the presence of HBr, providing stereoselective access to homoallylic bromides. It is also used in the synthesis of eta(2)-enonenickel complexes by reacting with [Ni(cod)(2)] (cod = 1,5-cyclooctadiene) and PBu(3) and to prepare a cyclopropylidene tetralone which underwent an enantioselective palladium-catalyzed [3+2] TMM cycloaddition providing a chiral spiro-fused cyclopentene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclopropanecarboxaldehyde reacts with Grignard or organolithium reagents to give the expected secondary alcohols. These are susceptible to ring opening in the presence of HBr, providing stereoselective access to homoallylic bromides. It is also used in the synthesis of eta(2)-enonenickel complexes by reacting with [Ni(cod)(2)] (cod = 1,5-cyclooctadiene) and PBu(3) and to prepare a cyclopropylidene tetralone which underwent an enantioselective palladium-catalyzed [3+2] TMM cycloaddition providing a chiral spiro-fused cyclopentene.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Recommended storage temperature is 2 - 8°C. It is sensitive to air.

Chemical Identifiers

CAS 1489-69-6
Molecular Formula C4H6O
Molecular Weight (g/mol) 70.091
MDL Number MFCD00012261
InChI Key JMYVMOUINOAAPA-UHFFFAOYSA-N
Synonym cyclopropanecarboxaldehyde, formylcyclopropane, cyclopropylmethanal, cyclopropanecarboxyldehyde, cyclopropanal, cyclopropane aldehyde, cyclopropancarbaldehyd, cyclopropancarbaldehyde, cyclopropancarboxaldehyde, cyclopropanecarboaldehyde
PubChem CID 123114
IUPAC Name cyclopropanecarbaldehyde
SMILES C1CC1C=O

Specifications

Density 0.938
Boiling Point 98°C to 101°C
Flash Point 7°C (44°F)
Odor Stench
Refractive Index 1.4298
Quantity 1 g
UN Number UN2924
Beilstein 906781
Sensitivity Air Sensitive
Solubility Information Soluble in water.
Formula Weight 70.09
Percent Purity 98%
Chemical Name or Material Cyclopropanecarboxaldehyde
Show More Show Less
Hazard Category H225-H314-H335
Hazard Statement GHS H Statement
H225-H314-H318
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
EINECSNumber 000-000-0
RTECSNumber GZ1005000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only