missing translation for 'onlineSavingsMsg'
Learn More

Cyclopropanecarbonyl chloride, 98%

Catalog No. AAA1194814
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1194814 25 g
AAA1194822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1194814 Supplier Thermo Scientific Chemicals Supplier No. A1194814
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 4023-34-1 | C4H5ClO | 104.533 g/mol

It is primarily used an intermediate in the manufacture of pharmaceutical and agrochemical products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is primarily used an intermediate in the manufacture of pharmaceutical and agrochemical products.

Solubility
Soluble in water (reacts violently).

Notes
Moisture Sensitive. Store away from oxidizing agents, bases, active metals and water/moisture. Store under inert gas.

Chemical Identifiers

CAS 4023-34-1
Molecular Formula C4H5ClO
Molecular Weight (g/mol) 104.533
MDL Number MFCD00001277
InChI Key ZOOSILUVXHVRJE-UHFFFAOYSA-N
Synonym cyclopropanecarboxylic acid chloride, cyclopropane carbonyl chloride, cyclopropylcarboxyl chloride, chloroformyl cyclopropane, cyclopropylcarbonyl chloride, chlorocarbonyl cyclopropane, cyclopropyl carbonyl chloride, cyclopropyl carboxylic chloride, cyclopropane carboxylic acid chloride, cyclopropanoyl chloride
PubChem CID 77637
IUPAC Name cyclopropanecarbonyl chloride
SMILES C1CC1C(=O)Cl

Specifications

Density 1.15
Boiling Point 119°C to 120°C
Flash Point 23°C (73°F)
Odor Pungent
Refractive Index 1.452
Quantity 25 g
UN Number UN2920
Beilstein 471286
Sensitivity Moisture sensitive
Solubility Information Soluble in water (reacts violently).
Formula Weight 104.54
Percent Purity 98%
Chemical Name or Material Cyclopropanecarbonyl chloride
Show More Show Less
Hazard Category H226-H301-H314
Hazard Statement GHS H Statement
H301-H314-H318-H226
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
Precautionary Statement P210-P233-P240-P241-P242-P243-P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, FLAMMABLE, N.O.S.
EINECSNumber 223-684-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only