missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclopropanecarbonitrile, 98%
Description
Cyclopropanecarbonitrile is used to prepare cis and trans crotonitrile and allyl cyanide. Further, it reacts with phenylmagnesium bromide to give cyclopropyl-phenyl ketone-imine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Cyclopropanecarbonitrile is used to prepare cis and trans crotonitrile and allyl cyanide. Further, it reacts with phenylmagnesium bromide to give cyclopropyl-phenyl ketone-imine.
Solubility
Miscible with water.
Notes
Incompatible with strong acids, strong bases, strong oxidizing agents and strong reducing agents.
Cyclopropanecarbonitrile is used to prepare cis and trans crotonitrile and allyl cyanide. Further, it reacts with phenylmagnesium bromide to give cyclopropyl-phenyl ketone-imine.
Solubility
Miscible with water.
Notes
Incompatible with strong acids, strong bases, strong oxidizing agents and strong reducing agents.
Specifications
Specifications
| Density | 0.905 |
| Boiling Point | 134°C to 135°C |
| Flash Point | 32°C (89°F) |
| Odor | Recognizable |
| Refractive Index | 1.421 |
| Quantity | 10 g |
| UN Number | UN3275 |
| Beilstein | 635790 |
| Solubility Information | Miscible with water. |
| Formula Weight | 67.09 |
| Show More |
Safety and Handling
| missing translation for 'healthHazard1' | H226-H301+H311+H331-H315-H319-H335 |
| missing translation for 'healthHazard2' | GHS H Statement H300-H310-H330-H226 Fatal if swallowed. Fatal in contact with skin. Fatal if inhaled. Flammable liquid and vapor. |
| missing translation for 'healthHazard3' | P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P311-P312-P330-P332+P313-P363-P370+P378q-P501c |
| DOTInformation | Hazard Class: 3; Packaging Group: III |
| EINECSNumber | 226-836-8 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only