missing translation for 'onlineSavingsMsg'
Learn More

Cyclopiazonic acid, 98%

Catalog No. AAJ61594MA
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ61594MA 10 mg
AAJ61594MF 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ61594MA Supplier Thermo Scientific Chemicals Supplier No. J61594MA

Inhibitor of the sarco/endoplasmic reticulum calcium pump, calcium-ATPase | CAS: 18172-33-3 | C20H20N2O3 | 336.391 g/mol

The use of Cyclopiazonic acid on Langendorff-perfused murine hearts displayed anti-arrhythmic effects. Also, administration of cyclopiazonic acid on rat soleus muscle showed a decrease in hormone sensitive lipase activity and triacylglycerol hydrolysis. It is also used as an inhibitor of endoplasmic reticulum Ca2+-ATPase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
The use of Cyclopiazonic acid on Langendorff-perfused murine hearts displayed anti-arrhythmic effects. Also, administration of cyclopiazonic acid on rat soleus muscle showed a decrease in hormone sensitive lipase activity and triacylglycerol hydrolysis. It is also used as an inhibitor of endoplasmic reticulum Ca2+-ATPase.

Solubility
Soluble in DMSO (100 mM), ethanol (5 mM), DMF (∼20 mg/ml), and 1:1 DMSO:PBS(pH 7.2) (0.5 mg/ml). Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: -20°C Keep in a dry place. Keep away from strong oxidizing agents.

Chemical Identifiers

CAS 18172-33-3
Molecular Formula C20H20N2O3
Molecular Weight (g/mol) 336.391
MDL Number MFCD00167445
InChI Key SZINUGQCTHLQAZ-AFAVFJNCSA-N
Synonym cyclopiazonic acid, alpha-cyclopiazonic acid, unii-x9tly4580z, ccris 4942, alpha-cyclopiazonate, cyclopyazonic acid, 6ar,10z,11as,11br-10-1-hydroxyethylidene-7,7-dimethyl-6a,7,11a,11b-tetrahydro-6h-pyrrolo 1',2':2,3 isoindolo 4,5,6-cd indole-9,11 2h,10h-dione, 9h-pyrrolo 1',2':2,3 isoindolo 4,5,6-cd indol-9-one, 2,6,6a,7,11a,11b-hexahydro-10-acetyl-7,7-dimethyl-11-hydroxy-, 6a-alpha,11a,11b-alpha, 6ar,11as,11br-10-acetyl-9-hydroxy-7,7-dimethyl-2,6,6a,7,11a,11b-hexahydro-11h-pyrrolo 1',2':2,3 isoindolo 4,5,6-cd indol-11-one, ambotzls-1020
PubChem CID 54711281
SMILES CC(=O)C1=C(C2C3C(CC4=C5C3=CNC5=CC=C4)C(N2C1=O)(C)C)O

Specifications

Color White
Quantity 10 mg
UN Number UN2811
Solubility Information Soluble in DMSO (100 mM),ethanol (5 mM),DMF (∽20mg/ml),and 1:1 DMSO:PBS(pH 7.2) (0.5mg/ml). Insoluble in water.
Formula Weight 336.39
Percent Purity ≥99%
Physical Form Powder
Chemical Name or Material Cyclopiazonic acid
Hazard Category H300
Hazard Statement GHS H Statement
H300-H361
Fatal if swallowed.
Suspected of damaging fertility or the unborn child.
Precautionary Statement P264b-P270-P301+P310-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
RTECSNumber UY8587000
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only