accessibility menu, dialog, popup

UserName

CH3OOHN(S)(R)(R)HNH3CCH3O

CAS RN 18172-33-3

IUPAC Name: (2R,3S,9R)-5-acetyl-6-hydroxy-8,8-dimethyl-7,16-diazapentacyclo[9.6.1.0²,⁹.0³,⁷.0¹⁵,¹⁸]octadeca-1(17),5,11(18),12,14-pentaen-4-one
Synonyms: cyclopiazonic acid
Molecular Weight (g/mol): 336.39
Molecular Formula: C20H20N2O3
InChi Key: RLOAZVAJNNPPDI-DQYPLSBCSA-N
SMILES: CC(=O)C1=C(O)N2[C@@H]([C@@H]3[C@@H](CC4=C5C(NC=C35)=CC=C4)C2(C)C)C1=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results