missing translation for 'onlineSavingsMsg'
Learn More

Cyclophosphamide Monohydrate 98.0+%, TCI America™

Catalog No. C223625G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C223625G 25 g
C22365G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C223625G Supplier TCI America Supplier No. C223625G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6055-19-2
Molecular Formula C7H17Cl2N2O3P
Molecular Weight (g/mol) 279.10
MDL Number MFCD00149395
InChI Key PWOQRKCAHTVFLB-UHFFFAOYNA-N
Synonym cyclophosphamide monohydrate, cyclophosphamide hydrate, cytoxan, endoxon, endoxan monohydrate, ciclophosphamide hydrat, cytoxan hydrate, endoxan a, cyclophosphamide hydrated
PubChem CID 22420
ChEBI CHEBI:4026
IUPAC Name 2-[bis(2-chloroethyl)amino]-1,3,2λ⁵-oxazaphosphinan-2-one hydrate
SMILES O.ClCCN(CCCl)P1(=O)NCCCO1

Specifications

Melting Point 51°C
Color White
Quantity 25 g
UN Number 3464
Formula Weight 261.08
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material Cyclophosphamide Monohydrate
RTECSNumber RP6157750
TSCA No
Recommended Storage Refrigerator