missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Cyclophilin A Monoclonal Antibody (C10)

Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA566053
Description
Cyclophilin A Monoclonal Antibody for Western Blot, ICC/IF, IHC
Immunophilins are a family of soluble cytosolic receptors capable of binding to one of two major immunosuppressant agents - cyclosporin A (CsA) or FK506. Proteins that bind FK506 are termed FK506 Binding Proteins (FKBPs) and those that bind cyclosporin A are called cyclophilins (CyP). Both CyP:CsA and FKBP:FK506 complexes have been shown to inhibit calcineurin, a calcium and calmodulin dependent protein phosphatase which has been implicated as an important signaling enzyme in T-cell activation, providing a possible mechanism of immunosuppression by CsA and FK506. Immunophilins function as peptidyl prolyl cis-trans-isomerases (PPIase) whose activity is inhibited by their respective immunosuppressant compounds. As PPIase's, immunophilins accelerate folding of some proteins both in vivo and in vitro by catalyzing slow steps in the initial folding and rearrangement of proline containing proteins. Within the cyclophilin family, there are several different proteins which show a high degree of homology including CyPA, CyPB and CyPC. CyPA, also termed CyP-18, is the most abundant and ubiquitous cyclophilin found in all vertebrate tissues and is present in T-cells.Chemical Identifiers
| Cyclophilin A | |
| 1 mg/mL | |
| Immunohistochemistry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| RUO | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| P10111 | |
| 25518 | |
| Recombinant Cyclophilin A (CYPA) (Val2-Leu164). | |
| Antibody |
| C10 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Monoclonal | |
| Liquid | |
| IgG1 κ | |
| Rat | |
| Ppia | |
| 2700098C05; Cphn; CYCA; cyclophilin; Cyclophilin A; Cyclosporin A-binding protein; CyP A; CyP-18; CYPA; CyP-A; CYPH; EC 5.2.1.8; epididymis secretory sperm binding protein Li 69p; HEL-S-69p; HGNC:9253; I79_011869; MGC117158; MGC12404; MGC23397; p1B15; p31; Peptidyl-prolyl cis-trans isomerase A; Peptidyl-prolyl cis-trans isomerase A, N-terminally processed; peptidylprolyl isomerase A; peptidylprolyl isomerase A (cyclophilin A); PPIA; PPIase; PPIase A; rotamase; rotamase A; SP18; T cell cyclophilin | |
| Ppia | |
| Primary | |
| Protein A/G |
Specifications
| Cyclophilin A | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| P10111 | |
| Ppia | |
| Recombinant Cyclophilin A (CYPA) (Val2-Leu164). | |
| 200 μL | |
| Primary | |
| Rat | |
| Antibody | |
| IgG1 κ |
| Immunohistochemistry, Western Blot, Immunocytochemistry | |
| C10 | |
| Unconjugated | |
| Ppia | |
| 2700098C05; Cphn; CYCA; cyclophilin; Cyclophilin A; Cyclosporin A-binding protein; CyP A; CyP-18; CYPA; CyP-A; CYPH; EC 5.2.1.8; epididymis secretory sperm binding protein Li 69p; HEL-S-69p; HGNC:9253; I79_011869; MGC117158; MGC12404; MGC23397; p1B15; p31; Peptidyl-prolyl cis-trans isomerase A; Peptidyl-prolyl cis-trans isomerase A, N-terminally processed; peptidylprolyl isomerase A; peptidylprolyl isomerase A (cyclophilin A); PPIA; PPIase; PPIase A; rotamase; rotamase A; SP18; T cell cyclophilin | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 25518 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |