missing translation for 'onlineSavingsMsg'
Learn More

Cyclopentyltrimethoxysilane, 95%

Catalog No. p-7046818
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1644214 25 g
AAL1644222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1644214 Supplier Thermo Scientific Chemicals Supplier No. L1644214
Only null left

CAS: 143487-47-2 | C8H18O3Si | 190.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 143487-47-2
Molecular Formula C8H18O3Si
Molecular Weight (g/mol) 190.31
MDL Number MFCD00236509
InChI Key YRMPTIHEUZLTDO-UHFFFAOYSA-N
Synonym acmc-20albx, cyclopentyltrimethoxysilan, cyclopentyl trimethoxysilane, silane, cyclopentyltrimethoxy-9ci
PubChem CID 9855701
SMILES CO[Si](OC)(OC)C1CCCC1

Specifications

Boiling Point 75°C (10mmHg)
Quantity 25 g
UN Number UN1993
Beilstein 8310738
Flash Point 54°C (129°F)
IUPAC Name cyclopentyl(trimethoxy)silane
Formula Weight 190.32
Percent Purity 97%
Refractive Index 1.425
Sensitivity Moisture sensitive
Density 0.99
Chemical Name or Material Cyclopentyltrimethoxysilane
Show More Show Less
Hazard Category H226-H315-H319
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P240-P241-P242-P243-P264b-P280-P303+P361+P353-P305+P351+P338-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only